The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136478.3 | NP_001129950.1 | 1 - 2 | 422 | 128 |
NM_001280559.1 | NP_001267488.1 | 1 - 2 | 422 | 128 | |
NM_001280560.1 | NP_001267489.1 | 1 - 2 | 422 | 128 | |
NM_030920.4 | NP_112182.1 | 1 - 2 | 422 | 128 | |
XM_005245513.3 | XP_005245570.1 | 1 - 2 | 420 | 128 | |
XM_005245514.3 | XP_005245571.1 | 1 - 2 | 420 | 128 | |
XM_017002418.1 | XP_016857907.1 | 1 - 2 | 420 | 128 | |
GenBank mRNA | AK092672.1 | - | 1 - 2 | 383 | 128 |
AK223469.1 | - | 1 - 2 | 289 | 128 | |
AK303333.1 | - | 1 - 2 | 222 | 128 | |
AK303392.1 | - | 1 - 2 | 288 | 128 | |
AL832664.1 | - | 1 - 2 | 288 | 128 | |
AL832674.1 | - | 1 - 2 | 424 | 128 | |
AL833660.1 | - | 1 - 2 | 176 | 128 | |
AY057381.1 | - | 1 - 2 | 361 | 128 | |
BC003380.1 | - | 1 - 2 | 241 | 128 | |
DA379137.1 | - | 1 - 2 | 422 | 128 |