The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282476.1 | NP_001269405.1 | 11 - 12 | 1728 | 119 |
NM_006380.3 | NP_006371.2 | 12 - 13 | 1817 | 119 | |
XM_011524198.2 | XP_011522500.1 | 13 - 14 | 1544 | 119 | |
XM_011524199.2 | XP_011522501.1 | 9 - 10 | 1029 | 119 | |
GenBank mRNA | AF017782.1 | - | 12 - 13 | 1793 | 119 |
AK292227.1 | - | 12 - 13 | 1584 | 119 | |
AK307666.1 | - | 11 - 12 | 1622 | 119 | |
BC018121.1 | - | 12 - 13 | 1720 | 119 | |
D86981.1 | - | 12 - 13 | 1796 | 119 |