Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DLX4
- Entrez Gene ID
-
1748
- Gene Name
- distal-less homeobox 4
- Gene Aliases
-
BP1, DLX7, DLX8, DLX9, OFC15
- Chromosome Location
-
Chr.17: 49968970 - 49974959 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U31762.1 |
- |
1 - 2 |
608 |
62 |
View Full Product Details