The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001292001.1 | NP_001278930.1 | 3 - 4 | 597 | 90 |
NM_003131.3 | NP_003122.1 | 3 - 4 | 1410 | 90 | |
GenBank mRNA | AK300467.1 | - | 3 - 4 | 597 | 90 |
BC048211.1 | - | 3 - 4 | 1246 | 90 | |
BC052572.1 | - | 3 - 4 | 1410 | 90 | |
J03161.1 | - | 3 - 4 | 1406 | 90 |