Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELL3
Entrez Gene ID
80237
Gene Name
elongation factor for RNA polymerase II 3
Gene Aliases
-
Chromosome Location
Chr.15: 43772600 - 43777304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025165.2 NP_079441.1 10 - 11 1490 90
GenBank mRNA AF276512.1 - 10 - 11 1087 90
AK026290.1 - 10 - 11 1172 90
AK057528.1 - 8 - 9 1175 90
AK126384.1 - 10 - 11 1729 90
BC006548.2 - 10 - 11 1141 90
BC019293.1 - 10 - 11 1141 90
CR457358.1 - 10 - 11 1087 90

View Full Product Details