Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
Chromosome Location
Chr.6: 31639028 - 31660900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098534.1 NP_001092004.1 21 - 22 3042 80
NM_001199698.1 NP_001186627.1 21 - 22 3172 80
NM_004639.3 NP_004630.3 21 - 22 3214 80
NM_080702.2 NP_542433.1 21 - 22 3172 80
NM_080703.2 NP_542434.1 21 - 22 3196 80
XM_011514892.2 XP_011513194.1 22 - 23 4296 80
XM_017011275.1 XP_016866764.1 22 - 23 4323 80
XM_017011276.1 XP_016866765.1 22 - 23 3313 80
XM_017011277.1 XP_016866766.1 22 - 23 3173 80
XM_017011278.1 XP_016866767.1 22 - 23 3248 80
XM_017011279.1 XP_016866768.1 22 - 23 3643 80
XM_017011280.1 XP_016866769.1 22 - 23 3215 80
XM_017011281.1 XP_016866770.1 22 - 23 3377 80
XM_017011282.1 XP_016866771.1 22 - 23 3169 80
XM_017011283.1 XP_016866772.1 22 - 23 4320 80
XM_017011284.1 XP_016866773.1 22 - 23 4290 80
XM_017011285.1 XP_016866774.1 21 - 22 3113 80
XM_017011286.1 XP_016866775.1 21 - 22 3132 80
XM_017011287.1 XP_016866776.1 22 - 23 4263 80
XM_017011288.1 XP_016866777.1 21 - 22 3050 80
XM_017011289.1 XP_016866778.1 21 - 22 4182 80
XM_017011291.1 XP_016866780.1 22 - 23 4323 80
XM_017011292.1 XP_016866781.1 21 - 22 4152 80
XM_017011293.1 XP_016866782.1 20 - 21 2945 80
XM_017011294.1 XP_016866783.1 20 - 21 4077 80
XM_017011296.1 XP_016866785.1 21 - 22 4152 80
XM_017011297.1 XP_016866786.1 21 - 22 3050 80
GenBank mRNA AK304879.1 - 21 - 22 3123 80
BC003133.1 - 21 - 22 3090 80
BX647244.1 - 21 - 22 3050 80
M33519.1 - 21 - 22 3149 80

View Full Product Details