Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
Chromosome Location
Chr.6: 31639028 - 31660900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098534.1 NP_001092004.1 9 - 10 1270 106
NM_001199697.1 NP_001186626.1 9 - 10 1400 106
NM_001199698.1 NP_001186627.1 9 - 10 1400 106
NM_004639.3 NP_004630.3 9 - 10 1442 106
NM_080702.2 NP_542433.1 9 - 10 1400 106
NM_080703.2 NP_542434.1 9 - 10 1424 106
XM_011514892.2 XP_011513194.1 9 - 10 2383 106
XM_017011275.1 XP_016866764.1 9 - 10 2383 106
XM_017011276.1 XP_016866765.1 9 - 10 1373 106
XM_017011277.1 XP_016866766.1 9 - 10 1233 106
XM_017011278.1 XP_016866767.1 9 - 10 1308 106
XM_017011279.1 XP_016866768.1 9 - 10 1703 106
XM_017011280.1 XP_016866769.1 9 - 10 1275 106
XM_017011281.1 XP_016866770.1 9 - 10 1437 106
XM_017011282.1 XP_016866771.1 9 - 10 1229 106
XM_017011283.1 XP_016866772.1 9 - 10 2383 106
XM_017011284.1 XP_016866773.1 9 - 10 2383 106
XM_017011285.1 XP_016866774.1 8 - 9 1173 106
XM_017011286.1 XP_016866775.1 8 - 9 1192 106
XM_017011287.1 XP_016866776.1 9 - 10 2383 106
XM_017011288.1 XP_016866777.1 8 - 9 1173 106
XM_017011289.1 XP_016866778.1 9 - 10 2383 106
XM_017011290.1 XP_016866779.1 9 - 10 2383 106
XM_017011291.1 XP_016866780.1 9 - 10 2383 106
XM_017011292.1 XP_016866781.1 9 - 10 2383 106
XM_017011293.1 XP_016866782.1 8 - 9 1173 106
XM_017011294.1 XP_016866783.1 9 - 10 2383 106
XM_017011295.1 XP_016866784.1 9 - 10 2383 106
XM_017011296.1 XP_016866785.1 9 - 10 2383 106
XM_017011297.1 XP_016866786.1 8 - 9 1173 106
XM_017011298.1 XP_016866787.1 9 - 10 2383 106
GenBank mRNA AK302695.1 - 9 - 10 1351 106
AK304879.1 - 9 - 10 1351 106
BC003133.1 - 9 - 10 1318 106
BX647244.1 - 9 - 10 1278 106
M33519.1 - 9 - 10 1377 106

View Full Product Details