Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCA5L
Entrez Gene ID
150082
Gene Name
LCA5L, lebercilin like
Gene Aliases
C21orf13
Chromosome Location
Chr.21: 39405844 - 39445796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152505.3 NP_689718.1 9 - 10 1598 99
XM_005260926.1 XP_005260983.1 9 - 10 1556 99
XM_006723967.1 XP_006724030.1 8 - 9 1601 99
XM_006723972.1 XP_006724035.1 9 - 10 1297 99
XM_006723973.1 XP_006724036.1 8 - 9 1137 99
XM_011529458.1 XP_011527760.1 11 - 12 2154 99
XM_011529459.1 XP_011527761.1 11 - 12 1813 99
XM_011529460.1 XP_011527762.1 12 - 13 1953 99
XM_011529461.1 XP_011527763.1 12 - 13 2060 99
XM_011529462.1 XP_011527764.1 10 - 11 1703 99
XM_011529463.2 XP_011527765.1 10 - 11 1756 99
XM_011529464.1 XP_011527766.1 11 - 12 1782 99
XM_011529465.1 XP_011527767.1 10 - 11 1701 99
XM_011529466.1 XP_011527768.1 10 - 11 1939 99
XM_011529467.1 XP_011527769.1 11 - 12 2017 99
XM_011529468.1 XP_011527770.1 10 - 11 1659 99
XM_011529469.2 XP_011527771.1 10 - 11 2046 99
XM_011529470.1 XP_011527772.1 9 - 10 1542 99
XM_011529471.1 XP_011527773.1 8 - 9 1610 99
XM_017028274.1 XP_016883763.1 9 - 10 1517 99
XM_017028275.1 XP_016883764.1 8 - 9 1270 99
XM_017028276.1 XP_016883765.1 9 - 10 1507 99
XM_017028277.1 XP_016883766.1 8 - 9 1190 99
XM_017028278.1 XP_016883767.1 7 - 8 1062 99
GenBank mRNA BC031059.2 - 9 - 10 1599 99
BC043006.1 - 9 - 10 1815 99
BX648744.1 - 6 - 7 1226 99

View Full Product Details