The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000629.2 | NP_000620.2 | 4 - 5 | 670 | 172 |
XM_005260964.2 | XP_005261021.1 | 4 - 5 | 643 | 172 | |
XM_011529552.1 | XP_011527854.1 | 4 - 5 | 670 | 172 | |
GenBank mRNA | AK222770.1 | - | 4 - 5 | 606 | 172 |
AK222812.1 | - | 4 - 5 | 606 | 172 | |
AK298051.1 | - | 4 - 5 | 657 | 172 | |
AK312631.1 | - | 4 - 5 | 606 | 172 | |
BC002590.2 | - | 4 - 5 | 585 | 172 | |
BC021825.1 | - | 4 - 5 | 602 | 172 | |
J03171.1 | - | 4 - 5 | 601 | 172 | |
JN543953.1 | - | 4 - 5 | 604 | 172 | |
JN543954.1 | - | 4 - 5 | 604 | 172 | |
JN543955.1 | - | 4 - 5 | 604 | 172 | |
JN543956.1 | - | 4 - 5 | 604 | 172 |