The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000629.2 | NP_000620.2 | 5 - 6 | 818 | 126 |
XM_005260964.2 | XP_005261021.1 | 5 - 6 | 791 | 126 | |
XM_011529552.1 | XP_011527854.1 | 5 - 6 | 818 | 126 | |
GenBank mRNA | AK222770.1 | - | 5 - 6 | 754 | 126 |
AK222812.1 | - | 5 - 6 | 754 | 126 | |
AK298051.1 | - | 5 - 6 | 805 | 126 | |
AK312631.1 | - | 5 - 6 | 754 | 126 | |
BC002590.2 | - | 5 - 6 | 733 | 126 | |
BC021825.1 | - | 5 - 6 | 750 | 126 | |
J03171.1 | - | 5 - 6 | 749 | 126 | |
JN543953.1 | - | 5 - 6 | 752 | 126 | |
JN543954.1 | - | 5 - 6 | 752 | 126 | |
JN543956.1 | - | 5 - 6 | 752 | 126 |