The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012086.4 | NP_036218.1 | 10 - 11 | 1543 | 89 |
XM_005246965.4 | XP_005247022.1 | 10 - 11 | 1547 | 89 | |
GenBank mRNA | AF133123.1 | - | 10 - 11 | 1479 | 89 |
AK025560.1 | - | 10 - 11 | 1457 | 89 | |
AK027050.1 | - | 4 - 5 | 494 | 89 | |
AK314718.1 | - | 10 - 11 | 1484 | 89 | |
AL137453.1 | - | 7 - 8 | 885 | 89 | |
BC015995.1 | - | 2 - 3 | 242 | 89 | |
BC043347.1 | - | 10 - 11 | 1398 | 89 |