The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024080.4 | NP_076985.4 | 18 - 19 | 2494 | 107 |
XM_011511810.2 | XP_011510112.1 | 18 - 19 | 2550 | 107 | |
XM_017004891.1 | XP_016860380.1 | 17 - 18 | 4361 | 107 | |
GenBank mRNA | AB061779.1 | - | 17 - 18 | 2427 | 107 |
AK303811.1 | - | 1 - 2 | 81 | 107 | |
AY090109.1 | - | 18 - 19 | 2494 | 107 | |
AY328400.1 | - | 17 - 18 | 2454 | 107 | |
BC126342.1 | - | 17 - 18 | 2414 | 107 | |
BC143819.1 | - | 17 - 18 | 2414 | 107 | |
DQ139309.1 | - | 17 - 18 | 2454 | 107 | |
KC692993.1 | - | 18 - 19 | 2521 | 107 | |
KC692994.1 | - | 4 - 5 | 593 | 107 | |
KC692995.1 | - | 3 - 4 | 480 | 107 |