Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEI1
Entrez Gene ID
150365
Gene Name
meiotic double-stranded break formation protein 1
Gene Aliases
SPATA38
Chromosome Location
Chr.22: 41699503 - 41799456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152513.3 NP_689726.3 22 - 23 2841 76
XM_011529935.1 XP_011528237.1 23 - 24 2814 76
XM_011529936.1 XP_011528238.1 22 - 23 2850 76
XM_011529938.1 XP_011528240.1 21 - 22 2733 76
XM_011529939.1 XP_011528241.1 22 - 23 2850 76
XM_011529941.1 XP_011528243.1 22 - 23 2850 76
XM_011529942.2 XP_011528244.1 22 - 23 3041 76
XM_011529943.1 XP_011528245.1 21 - 22 2574 76
XM_011529944.1 XP_011528246.1 22 - 23 2850 76
XM_011529945.2 XP_011528247.1 22 - 23 2852 76
XM_011529946.1 XP_011528248.1 19 - 20 2418 76
XM_011529947.1 XP_011528249.1 17 - 18 2328 76
XM_011529948.2 XP_011528250.1 18 - 19 2222 76
XM_011529949.1 XP_011528251.1 17 - 18 2159 76
XM_011529952.1 XP_011528254.1 17 - 18 2296 76
XM_011529954.1 XP_011528256.1 12 - 13 1600 76
XM_017028633.1 XP_016884122.1 22 - 23 2850 76
XM_017028635.1 XP_016884124.1 8 - 9 1180 76
GenBank mRNA AK092934.1 - 8 - 9 1210 76
AK093341.1 - 2 - 3 346 76
AK094461.1 - 4 - 5 627 76
AK128158.1 - 6 - 7 5565 76
AK301435.1 - 7 - 8 1047 76
AY952376.1 - 7 - 8 1553 76
BC029582.1 - 3 - 4 590 76

View Full Product Details