Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOS1
Entrez Gene ID
4842
Gene Name
nitric oxide synthase 1
Gene Aliases
IHPS1, N-NOS, NC-NOS, NOS, bNOS, nNOS
Chromosome Location
Chr.12: 117208142 - 117441843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000620.4 NP_000611.1 23 - 24 4331 94
NM_001204213.1 NP_001191142.1 22 - 23 2923 94
NM_001204214.1 NP_001191143.1 22 - 23 2918 94
NM_001204218.1 NP_001191147.1 24 - 25 4433 94
XM_011538398.2 XP_011536700.1 25 - 26 4304 94
XM_017019345.1 XP_016874834.1 24 - 25 4440 94
XM_017019346.1 XP_016874835.1 24 - 25 4254 94
XM_017019347.1 XP_016874836.1 23 - 24 4241 94
GenBank mRNA D16408.1 - 22 - 23 3620 94
EU753241.1 - 23 - 24 4305 94
L02881.1 - 22 - 23 4047 94
U17327.1 - 23 - 24 4305 94
U31466.1 - 22 - 23 3627 94

View Full Product Details