Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOS1
Entrez Gene ID
4842
Gene Name
nitric oxide synthase 1
Gene Aliases
IHPS1, N-NOS, NC-NOS, NOS, bNOS, nNOS
Chromosome Location
Chr.12: 117208142 - 117441843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000620.4 NP_000611.1 11 - 12 2649 70
NM_001204213.1 NP_001191142.1 10 - 11 1241 70
NM_001204214.1 NP_001191143.1 10 - 11 1236 70
NM_001204218.1 NP_001191147.1 11 - 12 2649 70
XM_011538398.2 XP_011536700.1 12 - 13 2520 70
XM_017019345.1 XP_016874834.1 11 - 12 2656 70
XM_017019346.1 XP_016874835.1 11 - 12 2470 70
XM_017019347.1 XP_016874836.1 11 - 12 2559 70
GenBank mRNA AK294435.1 - 11 - 12 2453 70
AK307481.1 - 11 - 12 2649 70
D16408.1 - 10 - 11 1938 70
EU753241.1 - 11 - 12 2623 70
L02881.1 - 10 - 11 2365 70
U17327.1 - 11 - 12 2623 70
U31466.1 - 10 - 11 1945 70

View Full Product Details