Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AACS
Entrez Gene ID
65985
Gene Name
acetoacetyl-CoA synthetase
Gene Aliases
ACSF1, SUR-5
Chromosome Location
Chr.12: 125065367 - 125143333 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK022740.1 - 8 - 9 884 95

View Full Product Details