The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319839.1 | NP_001306768.1 | 1 - 2 | 352 | 98 |
NM_001319840.1 | NP_001306769.1 | 1 - 2 | 352 | 98 | |
NM_023928.4 | NP_076417.2 | 1 - 2 | 352 | 98 | |
GenBank mRNA | AB054121.1 | - | 1 - 2 | 241 | 98 |
AK022451.1 | - | 1 - 2 | 282 | 98 | |
AK024036.1 | - | 1 - 2 | 281 | 98 | |
AK304176.1 | - | 1 - 2 | 279 | 98 | |
AK308779.1 | - | 1 - 2 | 285 | 98 | |
AY091468.1 | - | 1 - 2 | 113 | 98 | |
BC040490.1 | - | 1 - 2 | 340 | 98 | |
BC041000.1 | - | 1 - 2 | 335 | 98 | |
BC051862.1 | - | 1 - 2 | 258 | 98 |