The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001131065.1 | NP_001124537.1 | 1 - 2 | 1188 | 154 |
NM_173362.3 | NP_775498.1 | 1 - 2 | 1188 | 154 | |
GenBank mRNA | BC035110.2 | - | 1 - 2 | 1186 | 154 |
BC048979.1 | - | 1 - 2 | 1213 | 154 |