The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164160.1 | NP_001157632.1 | 15 - 16 | 1905 | 68 |
NM_001164161.1 | NP_001157633.1 | 15 - 16 | 1905 | 68 | |
XM_006718608.3 | XP_006718671.1 | 16 - 17 | 2021 | 68 | |
XM_006718613.2 | XP_006718676.1 | 16 - 17 | 2021 | 68 | |
XM_011545135.1 | XP_011543437.1 | 16 - 17 | 2021 | 68 | |
XM_011545136.2 | XP_011543438.1 | 15 - 16 | 1904 | 68 | |
XM_011545137.2 | XP_011543439.1 | 16 - 17 | 2004 | 68 | |
XM_011545138.2 | XP_011543440.1 | 17 - 18 | 2222 | 68 | |
XM_011545140.2 | XP_011543442.1 | 16 - 17 | 2021 | 68 | |
XM_011545145.2 | XP_011543447.1 | 15 - 16 | 1868 | 68 | |
XM_017017962.1 | XP_016873451.1 | 15 - 16 | 1859 | 68 | |
XM_017017963.1 | XP_016873452.1 | 16 - 17 | 1945 | 68 | |
XM_017017977.1 | XP_016873466.1 | 15 - 16 | 1792 | 68 | |
XM_017017978.1 | XP_016873467.1 | 14 - 15 | 1643 | 68 | |
XM_017017980.1 | XP_016873469.1 | 14 - 15 | 1643 | 68 | |
XM_017017983.1 | XP_016873472.1 | 15 - 16 | 2042 | 68 | |
GenBank mRNA | AB046778.2 | - | 15 - 16 | 1853 | 68 |
AK057250.1 | - | 9 - 10 | 978 | 68 | |
AK303315.1 | - | 10 - 11 | 1221 | 68 | |
BC105933.1 | - | 14 - 15 | 1773 | 68 | |
BC105934.1 | - | 14 - 15 | 1773 | 68 | |
CR933658.1 | - | 15 - 16 | 1868 | 68 | |
CR936718.1 | - | 15 - 16 | 1873 | 68 | |
DQ111954.2 | - | 13 - 14 | 1638 | 68 |