The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024838.4 | NP_079114.3 | 1 - 2 | 115 | 116 |
GenBank mRNA | DA496769.1 | - | 1 - 2 | 128 | 116 |