The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015638.2 | NP_056453.1 | 1 - 2 | 199 | 111 |
NM_199368.1 | NP_955400.1 | 1 - 2 | 199 | 111 | |
XM_011528772.2 | XP_011527074.1 | 1 - 2 | 221 | 111 | |
XM_017027799.1 | XP_016883288.1 | 1 - 2 | 221 | 111 | |
XM_017027800.1 | XP_016883289.1 | 1 - 2 | 221 | 111 | |
GenBank mRNA | AK074106.1 | - | 1 - 2 | 178 | 112 |
AK303476.1 | - | 1 - 2 | 191 | 111 | |
AK315582.1 | - | 1 - 2 | 191 | 111 | |
BC013144.1 | - | 1 - 2 | 168 | 111 | |
BI488542.1 | - | 1 - 2 | 180 | 111 | |
BM149369.1 | - | 1 - 2 | 199 | 111 | |
CF125531.1 | - | 1 - 2 | 171 | 111 |