The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182658.2 | NP_872599.1 | 12 - 13 | 1358 | 75 |
GenBank mRNA | AF549189.1 | - | 12 - 13 | 1265 | 75 |
BC113398.1 | - | 12 - 13 | 1358 | 75 | |
BC113400.1 | - | 12 - 13 | 1358 | 75 |