The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182658.2 | NP_872599.1 | 3 - 4 | 479 | 88 |
GenBank mRNA | BC113398.1 | - | 3 - 4 | 479 | 88 |
BC113400.1 | - | 3 - 4 | 479 | 88 |