Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFB3
Entrez Gene ID
359710
Gene Name
BPI fold containing family B member 3
Gene Aliases
C20orf185, LPLUNC3, RYA3
Chromosome Location
Chr.20: 33055331 - 33073637 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182658.2 NP_872599.1 9 - 10 1069 96
GenBank mRNA AF549189.1 - 9 - 10 976 96
BC113398.1 - 9 - 10 1069 96
BC113400.1 - 9 - 10 1069 96

View Full Product Details