The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079673.1 | NP_001073141.1 | 1 - 2 | 270 | 80 |
XM_017020442.1 | XP_016875931.1 | 1 - 2 | 201 | 80 | |
GenBank mRNA | AJ749706.1 | - | 1 - 2 | 272 | 80 |
AK056071.1 | - | 1 - 2 | 254 | 80 | |
BX648141.1 | - | 1 - 2 | 260 | 80 | |
BX648706.1 | - | 1 - 2 | 268 | 80 |