The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025180.3 | NP_079456.2 | 14 - 15 | 2119 | 123 |
XM_005247795.4 | XP_005247852.1 | 13 - 14 | 1792 | 123 | |
XM_005247797.3 | XP_005247854.1 | 13 - 14 | 2047 | 123 | |
XM_005247799.3 | XP_005247856.1 | 13 - 14 | 1860 | 123 | |
XM_006713760.3 | XP_006713823.1 | 13 - 14 | 1724 | 123 | |
XM_011513194.2 | XP_011511496.1 | 13 - 14 | 1841 | 123 | |
XM_011513197.2 | XP_011511499.1 | 12 - 13 | 1675 | 123 | |
XM_017007244.1 | XP_016862733.1 | 13 - 14 | 1835 | 123 | |
XM_017007245.1 | XP_016862734.1 | 12 - 13 | 1694 | 123 | |
XM_017007246.1 | XP_016862735.1 | 12 - 13 | 1586 | 123 | |
XM_017007247.1 | XP_016862736.1 | 12 - 13 | 1703 | 123 | |
XM_017007248.1 | XP_016862737.1 | 12 - 13 | 1870 | 123 | |
XM_017007249.1 | XP_016862738.1 | 13 - 14 | 1959 | 123 | |
XM_017007250.1 | XP_016862739.1 | 13 - 14 | 1833 | 123 | |
XM_017007252.1 | XP_016862741.1 | 13 - 14 | 1766 | 123 | |
GenBank mRNA | AK056465.1 | - | 14 - 15 | 2097 | 123 |