The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020799.3 | NP_065850.1 | 5 - 6 | 925 | 150 |
XM_006717928.2 | XP_006717991.1 | 6 - 7 | 1026 | 150 | |
XM_006717930.3 | XP_006717993.1 | 5 - 6 | 1513 | 150 | |
XM_011539983.1 | XP_011538285.1 | 6 - 7 | 1433 | 150 | |
XM_011539985.1 | XP_011538287.1 | 6 - 7 | 756 | 150 | |
XM_017016453.1 | XP_016871942.1 | 6 - 7 | 1750 | 150 | |
XM_017016454.1 | XP_016871943.1 | 7 - 8 | 1234 | 150 | |
XM_017016455.1 | XP_016871944.1 | 8 - 9 | 1689 | 150 | |
XM_017016456.1 | XP_016871945.1 | 6 - 7 | 1346 | 150 | |
XM_017016457.1 | XP_016871946.1 | 5 - 6 | 1272 | 150 | |
XM_017016458.1 | XP_016871947.1 | 3 - 4 | 450 | 150 | |
GenBank mRNA | AB010120.1 | - | 4 - 5 | 453 | 150 |
AB037794.1 | - | 5 - 6 | 1246 | 150 | |
AK056086.1 | - | 5 - 6 | 854 | 150 | |
AK092946.1 | - | 4 - 5 | 1054 | 150 | |
BC010846.1 | - | 5 - 6 | 843 | 150 | |
BX649096.1 | - | 6 - 7 | 9348 | 150 |