The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006346.2 | NP_006337.2 | 6 - 7 | 1110 | 121 |
XM_011534881.2 | XP_011533183.1 | 6 - 7 | 1138 | 121 | |
XM_011534882.2 | XP_011533184.1 | 6 - 7 | 1138 | 121 | |
XM_011534884.2 | XP_011533186.1 | 6 - 7 | 1138 | 121 | |
XM_011534885.2 | XP_011533187.1 | 5 - 6 | 765 | 121 | |
XM_011534886.2 | XP_011533188.1 | 6 - 7 | 1138 | 121 | |
XM_017020350.1 | XP_016875839.1 | 5 - 6 | 764 | 121 | |
XM_017020351.1 | XP_016875840.1 | 6 - 7 | 1138 | 121 | |
XM_017020352.1 | XP_016875841.1 | 6 - 7 | 1064 | 121 | |
XM_017020353.1 | XP_016875842.1 | 6 - 7 | 1061 | 121 | |
XM_017020354.1 | XP_016875843.1 | 6 - 7 | 1061 | 121 | |
GenBank mRNA | AF330046.1 | - | 6 - 7 | 1152 | 121 |
AK309828.1 | - | 6 - 7 | 1067 | 121 | |
BC006009.1 | - | 6 - 7 | 1063 | 121 | |
BC008432.1 | - | 6 - 7 | 1063 | 121 | |
BC041806.1 | - | 6 - 7 | 1062 | 121 | |
BC051911.1 | - | 6 - 7 | 1040 | 121 |