The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020846.1 | NP_065897.1 | 5 - 6 | 1369 | 73 |
XM_005248121.2 | XP_005248178.1 | 5 - 6 | 1485 | 73 | |
GenBank mRNA | AB040891.1 | - | 5 - 6 | 1339 | 73 |
AK025264.1 | - | 1 - 2 | 420 | 73 | |
AK026476.1 | - | 5 - 6 | 916 | 73 | |
AK291006.1 | - | 5 - 6 | 1499 | 73 | |
BC031691.2 | - | 3 - 4 | 689 | 73 | |
BC040993.1 | - | 5 - 6 | 1648 | 73 |