Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMBN
Entrez Gene ID
258
Gene Name
ameloblastin
Gene Aliases
AI1F
Chromosome Location
Chr.4: 70592258 - 70607288 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016519.5 NP_057603.1 4 - 5 281 91
GenBank mRNA AF209780.1 - 4 - 5 263 91
AF219994.2 - 4 - 5 245 91
AF263464.1 - 4 - 5 255 91
AJ537437.1 - 4 - 5 255 91
AK314918.1 - 4 - 5 216 91
BC106931.1 - 4 - 5 216 91
BC106932.1 - 4 - 5 216 91
BP248666.1 - 4 - 5 281 91

View Full Product Details