The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007044.3 | NP_008975.1 | 5 - 6 | 971 | 162 |
XM_005266804.2 | XP_005266861.1 | 5 - 6 | 751 | 162 | |
XM_005266805.3 | XP_005266862.1 | 5 - 6 | 890 | 162 | |
XM_017010207.1 | XP_016865696.1 | 6 - 7 | 1061 | 162 | |
XM_017010210.1 | XP_016865699.1 | 5 - 6 | 756 | 162 | |
GenBank mRNA | AF056022.1 | - | 4 - 5 | 667 | 162 |
AK292090.1 | - | 5 - 6 | 881 | 162 |