Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHISA6
Entrez Gene ID
388336
Gene Name
shisa family member 6
Gene Aliases
-
Chromosome Location
Chr.17: 11241423 - 11564063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207386.3 NP_997269.2 4 - 5 952 71
XM_017024618.1 XP_016880107.1 3 - 4 856 71
GenBank mRNA AK128003.1 - 3 - 4 295 71

View Full Product Details