Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC73
Entrez Gene ID
493860
Gene Name
coiled-coil domain containing 73
Gene Aliases
NY-SAR-79
Chromosome Location
Chr.11: 32602080 - 32794658 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC062749.1 - 11 - 12 902 134

View Full Product Details