The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006093.3 | NP_006084.2 | 4 - 5 | 617 | 64 |
GenBank mRNA | AF132209.2 | - | 4 - 5 | 617 | 64 |
AY358930.1 | - | 4 - 5 | 597 | 64 | |
BC069126.1 | - | 4 - 5 | 614 | 64 | |
BC101613.1 | - | 3 - 4 | 556 | 64 | |
BC113411.1 | - | 3 - 4 | 556 | 64 |