The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320778.1 | NP_001307707.1 | 6 - 7 | 731 | 69 |
NM_005697.4 | NP_005688.2 | 5 - 6 | 602 | 69 | |
XM_017021856.1 | XP_016877345.1 | 6 - 7 | 656 | 69 | |
GenBank mRNA | AF005038.2 | - | 5 - 6 | 507 | 69 |
AK096376.1 | - | 4 - 5 | 493 | 69 | |
AK096794.1 | - | 4 - 5 | 2391 | 69 | |
AK291884.1 | - | 5 - 6 | 588 | 69 | |
AK315516.1 | - | 5 - 6 | 551 | 69 | |
BC001376.2 | - | 5 - 6 | 552 | 69 | |
BC004385.1 | - | 5 - 6 | 552 | 69 | |
CD300387.1 | - | 6 - 7 | 707 | 69 | |
CR456800.1 | - | 5 - 6 | 478 | 69 |