The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320778.1 | NP_001307707.1 | 7 - 8 | 886 | 113 |
NM_005697.4 | NP_005688.2 | 6 - 7 | 757 | 113 | |
XM_017021856.1 | XP_016877345.1 | 7 - 8 | 811 | 113 | |
GenBank mRNA | AF005038.2 | - | 6 - 7 | 662 | 113 |
AK096376.1 | - | 5 - 6 | 648 | 113 | |
AK096794.1 | - | 5 - 6 | 2546 | 113 | |
AK291884.1 | - | 6 - 7 | 743 | 113 | |
AK315516.1 | - | 6 - 7 | 706 | 113 | |
BC001376.2 | - | 6 - 7 | 707 | 113 | |
BC004385.1 | - | 6 - 7 | 707 | 113 | |
CR456800.1 | - | 6 - 7 | 633 | 113 |