The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166103.1 | NP_001159575.1 | 2 - 3 | 599 | 69 |
NM_021102.3 | NP_066925.1 | 3 - 4 | 770 | 69 | |
GenBank mRNA | AB006534.1 | - | 3 - 4 | 647 | 69 |
AF027205.1 | - | 3 - 4 | 699 | 69 | |
AK291532.1 | - | 3 - 4 | 585 | 69 | |
AK297154.1 | - | 2 - 3 | 274 | 69 | |
AX772830.1 | - | 2 - 3 | 481 | 69 | |
BC001668.2 | - | 3 - 4 | 666 | 69 | |
BC007705.2 | - | 3 - 4 | 614 | 69 | |
BC011951.1 | - | 3 - 4 | 570 | 69 | |
BC011955.1 | - | 3 - 4 | 542 | 69 | |
BC012868.2 | - | 3 - 4 | 657 | 69 | |
BT020115.1 | - | 3 - 4 | 335 | 69 | |
BT020116.1 | - | 3 - 4 | 335 | 69 | |
BT020117.1 | - | 3 - 4 | 335 | 69 | |
CR407604.1 | - | 3 - 4 | 335 | 69 | |
U78095.1 | - | 3 - 4 | 635 | 69 |