Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C12orf45
Entrez Gene ID
121053
Gene Name
chromosome 12 open reading frame 45
Gene Aliases
-
Chromosome Location
Chr.12: 104986320 - 104994727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152318.2 NP_689531.2 3 - 4 373 87
GenBank mRNA BC032326.1 - - 357 87
BG829346.1 - 3 - 4 374 87

View Full Product Details