The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000441.1 | NP_000432.1 | 16 - 17 | 2029 | 111 |
XM_005250425.2 | XP_005250482.1 | 16 - 17 | 2012 | 111 | |
XM_017012318.1 | XP_016867807.1 | 14 - 15 | 1730 | 111 | |
GenBank mRNA | AF030880.1 | - | 16 - 17 | 2029 | 111 |
AK294388.1 | - | 7 - 8 | 908 | 111 | |
AK298543.1 | - | 6 - 7 | 652 | 111 | |
AK300604.1 | - | 7 - 8 | 724 | 111 |