The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000441.1 | NP_000432.1 | 2 - 3 | 389 | 80 |
XM_005250425.2 | XP_005250482.1 | 2 - 3 | 372 | 80 | |
XM_006716025.3 | XP_006716088.1 | 1 - 2 | 168 | 80 | |
XM_017012318.1 | XP_016867807.1 | 1 - 2 | 168 | 80 | |
GenBank mRNA | AF030880.1 | - | 2 - 3 | 389 | 80 |