The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000441.1 | NP_000432.1 | 7 - 8 | 1137 | 122 |
XM_005250425.2 | XP_005250482.1 | 7 - 8 | 1120 | 122 | |
XM_006716025.3 | XP_006716088.1 | 6 - 7 | 916 | 122 | |
XM_017012318.1 | XP_016867807.1 | 6 - 7 | 916 | 122 | |
GenBank mRNA | AF030880.1 | - | 7 - 8 | 1137 | 122 |