Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC26A4
Entrez Gene ID
5172
Gene Name
solute carrier family 26 member 4
Gene Aliases
DFNB4, EVA, PDS, TDH2B
Chromosome Location
Chr.7: 107660635 - 107717809 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000441.1 NP_000432.1 8 - 9 1222 120
XM_005250425.2 XP_005250482.1 8 - 9 1205 120
XM_006716025.3 XP_006716088.1 7 - 8 1001 120
XM_017012318.1 XP_016867807.1 7 - 8 1001 120
GenBank mRNA AF030880.1 - 8 - 9 1222 120

View Full Product Details