The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256604.1 | NP_001243533.1 | 3 - 4 | 627 | 127 |
NM_001256605.1 | NP_001243534.1 | 2 - 3 | 369 | 127 | |
NM_001256608.1 | NP_001243537.1 | 3 - 4 | 627 | 127 | |
NM_001256609.1 | NP_001243538.1 | 3 - 4 | 856 | 127 | |
NM_144580.2 | NP_653181.1 | 3 - 4 | 627 | 127 | |
XM_011509117.2 | XP_011507419.1 | 2 - 3 | 286 | 127 | |
XM_017000149.1 | XP_016855638.1 | 3 - 4 | 623 | 127 | |
GenBank mRNA | AK075349.1 | - | 3 - 4 | 596 | 127 |
AK296157.1 | - | 3 - 4 | 824 | 127 | |
AY358450.1 | - | 3 - 4 | 600 | 127 | |
BC011575.1 | - | 3 - 4 | 610 | 127 | |
BC018757.1 | - | 3 - 4 | 596 | 127 | |
BC036340.1 | - | 3 - 4 | 543 | 127 | |
BP302818.1 | - | 1 - 2 | 94 | 127 | |
CX756681.1 | - | 3 - 4 | 542 | 127 |