Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPC5
Entrez Gene ID
7224
Gene Name
transient receptor potential cation channel subfamily C member 5
Gene Aliases
PPP1R159, TRP5
Chromosome Location
Chr.X: 111774314 - 112082943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012471.2 NP_036603.1 9 - 10 3061 112
XM_017029774.1 XP_016885263.1 10 - 11 3351 112
GenBank mRNA AF054568.1 - 9 - 10 3060 112
BC137271.1 - 9 - 10 2901 112
BC137274.1 - 9 - 10 2901 112

View Full Product Details