The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147164.2 | NP_671693.1 | 1 - 2 | 187 | 145 |
XM_017014260.1 | XP_016869749.1 | 1 - 2 | 159 | 145 | |
GenBank mRNA | M73238.1 | - | 1 - 2 | 156 | 145 |