Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN16
Entrez Gene ID
10686
Gene Name
claudin 16
Gene Aliases
HOMG3, PCLN1
Chromosome Location
Chr.3: 190387872 - 190412143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006580.3 NP_006571.1 1 - 2 565 61
GenBank mRNA AF152101.1 - 1 - 2 385 61
AK314955.1 - 1 - 2 564 61
BC069662.1 - 1 - 2 385 61
BC069682.1 - 1 - 2 385 61
BC069759.1 - 1 - 2 385 61
BC069777.1 - 1 - 2 385 61

View Full Product Details