Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35A5
Entrez Gene ID
55032
Gene Name
solute carrier family 35 member A5
Gene Aliases
-
Chromosome Location
Chr.3: 112561709 - 112584439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017945.3 NP_060415.1 1 - 2 135
GenBank mRNA AK000737.1 - 1 - 2 169 135
AK098839.1 - 1 - 2 135
AK297411.1 - 1 - 2 135
AY358994.1 - 1 - 2 135
BC005207.1 - 1 - 2 135
BC010307.1 - 1 - 2 135
BC013046.1 - 1 - 2 135
DA854399.1 - 1 - 2 135

View Full Product Details