The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033054.2 | NP_149043.2 | 13 - 14 | 1800 | 52 |
XM_017012503.1 | XP_016867992.1 | 12 - 13 | 1559 | 52 | |
XM_017012504.1 | XP_016867993.1 | 13 - 14 | 1795 | 52 | |
GenBank mRNA | AB290179.1 | - | 13 - 14 | 1760 | 52 |
AF380932.1 | - | 12 - 13 | 1560 | 52 | |
AF380933.1 | - | 12 - 13 | 1560 | 52 | |
AK074135.1 | - | 9 - 10 | 1573 | 52 | |
AK131089.1 | - | 8 - 9 | 3917 | 52 | |
AL832302.1 | - | 12 - 13 | 1555 | 52 | |
BC113544.1 | - | 13 - 14 | 1747 | 52 | |
BC117241.1 | - | 13 - 14 | 1747 | 52 | |
BI910371.1 | - | 5 - 6 | 498 | 52 |