Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO1G
Entrez Gene ID
64005
Gene Name
myosin IG
Gene Aliases
HA2, HLA-HA2, MHAG
Chromosome Location
Chr.7: 44962661 - 44979105 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033054.2 NP_149043.2 14 - 15 1924 55
XM_017012503.1 XP_016867992.1 13 - 14 1683 55
XM_017012504.1 XP_016867993.1 14 - 15 1919 55
GenBank mRNA AB290179.1 - 14 - 15 1884 55
AF380932.1 - 13 - 14 1684 55
AF380933.1 - 13 - 14 1684 55
AK074135.1 - 10 - 11 1697 55
AK131089.1 - 9 - 10 4041 55
AL832302.1 - 13 - 14 1679 55
BC113544.1 - 14 - 15 1871 55
BC117241.1 - 14 - 15 1871 55
BI910371.1 - 6 - 7 622 55

View Full Product Details