The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033054.2 | NP_149043.2 | 15 - 16 | 2095 | 66 |
XM_017012503.1 | XP_016867992.1 | 14 - 15 | 1854 | 66 | |
XM_017012504.1 | XP_016867993.1 | 15 - 16 | 2090 | 66 | |
GenBank mRNA | AB290179.1 | - | 15 - 16 | 2055 | 66 |
AK131089.1 | - | 10 - 11 | 4212 | 66 | |
AL832302.1 | - | 14 - 15 | 1850 | 66 | |
BC113544.1 | - | 15 - 16 | 2042 | 66 | |
BC117241.1 | - | 15 - 16 | 2042 | 66 |